missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromo-3-methylphenol, 98+%, Thermo Scientific™

Product Code. 11317276
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11317276

Brand: Thermo Scientific Alfa Aesar L09183.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4619-74-3
Molecular Formula C7H5Br3O
Molecular Weight (g/mol) 344.828
MDL Number MFCD00019976
InChI Key QKHROXOPRBWBDD-UHFFFAOYSA-N
Synonym micatex, triphysan, triphysol, 2,4,6-tribromo-m-cresol, tribromometacresol, m-cresol, 2,4,6-tribromo, phenol, 2,4,6-tribromo-3-methyl, 2,4,6-tribromo-m-cresol oh=1, acmc-20ah2c, 2,6-tribromo-m-cresol
PubChem CID 20737
IUPAC Name 2,4,6-tribromo-3-methylphenol
SMILES CC1=C(C(=C(C=C1Br)Br)O)Br

Specifications

Melting Point 81°C to 83°C
Quantity 100 g
Beilstein 1949974
Merck Index 14,9612
Formula Weight 344.84
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Tribromo-3-methylphenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.