missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromo-3-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11372636
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372636 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11372636 Supplier Thermo Scientific Alfa Aesar Supplier No. L08656.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2737-22-6 | Molecular Formula: C7H3Br3O2 | Molecular Weight: 358.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2737-22-6
Molecular Formula C7H3Br3O2
Molecular Weight (g/mol) 358.81
MDL Number MFCD00017320
InChI Key FAWOIFAFFUDNJX-UHFFFAOYSA-N
Synonym 3-hydroxy-2,4,6-tribromobenzaldehyde, benzaldehyde, 3-hydroxy-2,4,6-tribromo, benzaldehyde,2,4,6-tribromo-3-hydroxy, acmc-1cj0g, 3-08-00-00209 beilstein handbook reference, 2,4,6-tris bromanyl-3-oxidanyl-benzaldehyde
PubChem CID 200803
IUPAC Name 2,4,6-tribromo-3-hydroxybenzaldehyde
SMILES OC1=C(Br)C=C(Br)C(C=O)=C1Br

Specifications

Melting Point 119°C to 122°C
Quantity 10 g
Beilstein 2098037
Sensitivity Air Sensitive
Formula Weight 358.81
Percent Purity 98%
Chemical Name or Material 2,4,6-Tribromo-3-hydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.