missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triaminopyrimidine, 97%

Product Code. 10143140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10143140 100 g Plastic bottle 100g
10189831 5 g Glass Bottle 5g
10713901 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10143140 Supplier Thermo Scientific Acros Supplier No. 139251000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1004-38-2 | C4H7N5 | 125.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1004-38-2
MDL Number MFCD00006100
InChI Key JTTIOYHBNXDJOD-UHFFFAOYSA-N
Synonym 2,4,6-triaminopyrimidine, 2,4,6-pyrimidinetriamine, pyrimidine-2,4,6-triyltriamine, pyrimidine, 2,4,6-triamino, triamino pyrimidine, 3ay, aaat, pubchem7216, 2,6-pyrimidinetriamine, 2,6-triaminopyrimidine
PubChem CID 13863
ChEBI CHEBI:39857
IUPAC Name pyrimidine-2,4,6-triamine
SMILES C1=C(N=C(N=C1N)N)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 246°C to 252°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Molecular Formula C4H7N5
Quantity 100 g
Molecular Weight (g/mol) 125.135
Formula Weight 125.13
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 2, 4, 6-Triaminopyrimidine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,6-Triaminopyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.