missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri-tert-butylphenol, TRC

Product Code. 30137873
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137873 10 g 10g
18061550 5 g 5g
18080240 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137873 Supplier Toronto Research Chemicals Supplier No. TRCT07773510G
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 732-26-3
Molecular Formula C18 H30 O
Molecular Weight (g/mol) 262.43
Synonym Phenol, 2,4,6-tris(1,1-dimethylethyl)-, Phenol, 2,4,6-tri-tert-butyl- (6CI,7CI,8CI), 2,4,6-Tris(1,1-dimethylethyl)phenol, 2,4,6-Tri-t-butylphenol, 2,4,6-Tri-tert-butyl-1-hydroxybenzene, 2,4,6-Tri-tert-butylphenol, 2,4,6-Tris(tert-butyl)phenol, 2,4,6-Tritert-butylphenol, Alkofen B, Antioxidant 333, Antioxidant Q 46, NSC 14459, P 23, P 23 (phenol), TM 02, Tri-tert-butylphenol, Voidox
IUPAC Name 2,4,6-tritert-butylphenol
SMILES CC(C)(C)c1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C18H30O/c1-16(2,3)12-10-13(17(4,5)6)15(19)14(11-12)18(7,8)9/h10-11,19H,1-9H3
Formula Weight 262.2297
Percent Purity >95%
Chemical Name or Material 2,4,6-Tri-tert-butylphenol
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.