missing translation for 'onlineSavingsMsg'
Learn More

2-(4,6-Diamino-1,3,5-triazin-2-yl)sulfanylethanesulfonic Acid, TRC

Product Code. 30133858
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133858 250 mg 250mg
17942785 100 mg 100mg
17962595 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133858 Supplier Toronto Research Chemicals Supplier No. TRCD416770250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1391054-56-0
Molecular Formula C5 H9 N5 O3 S2
Molecular Weight (g/mol) 251.29
Synonym 2-[(4,6-Diamino-1,3,5-triazin-2-yl)thio]ethanesulfonic Acid, 2-(4,6-Diamino-1,3,5-triazin-2-yl)sulfanylethanesulfonic Acid, Mesna Imp. E (EP)
IUPAC Name 2-[(4,6-diamino-1,3,5-triazin-2-yl)sulfanyl]ethanesulfonic acid
SMILES Nc1nc(N)nc(SCCS(=O)(=O)O)n1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H9N5O3S2/c6-3-8-4(7)10-5(9-3)14-1-2-15(11,12)13/h1-2H2,(H,11,12,13)(H4,6,7,8,9,10)
Formula Weight 251.0147
Chemical Name or Material 2-(4,6-Diamino-1,3,5-triazin-2-yl)sulfanylethanesulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.