missing translation for 'onlineSavingsMsg'
Learn More

2,4,6,8-Tetrachloroquinazoline, 97%, Thermo Scientific™

Product Code. 15420969
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420969 1 g 1g
15430969 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420969 Supplier Thermo Scientific Alfa Aesar Supplier No. H63968.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,4,6,8-Tetrachloroquinazoline is employed as a intermediate for pharmaceutical and organic synthesis.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54185-42-1
Molecular Formula C8H2Cl4N2
Molecular Weight (g/mol) 267.918
MDL Number MFCD03844695
InChI Key GYUFSVVBAINEBI-UHFFFAOYSA-N
Synonym quinazoline, 2,4,6,8-tetrachloro
PubChem CID 12703363
IUPAC Name 2,4,6,8-tetrachloroquinazoline
SMILES C1=C(C=C2C(=C1Cl)N=C(N=C2Cl)Cl)Cl

Specifications

Quantity 1 g
Formula Weight 267.93
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,4,6,8-Tetrachloroquinazoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.