missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethyloxazole, 97%, Thermo Scientific Chemicals

Catalog Number 11474093
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
20662-84-4
C6H9NO
111.144
MFCD00005308
ZRLDBDZSLLGDOX-UHFFFAOYSA-N
2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636
30215
2,4,5-trimethyl-1,3-oxazole
CC1=C(OC(=N1)C)C
This item is not returnable. View return policy
20662-84-4
C6H9NO
111.144
MFCD00005308
ZRLDBDZSLLGDOX-UHFFFAOYSA-N
2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636
30215
2,4,5-trimethyl-1,3-oxazole
CC1=C(OC(=N1)C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.964
Boiling Point 133°C to 134°C
Flash Point 33°C (91°F)
Odor Pungent
Refractive Index 1.442
Quantity 5 g
UN Number UN1993
Beilstein 107043
Formula Weight 111.14
Percent Purity 97%
Chemical Name or Material 2,4,5-Trimethyloxazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'