missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzyl chloride, 97%, Thermo Scientific™

Catalog Number 11494447
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
243139-71-1
C7H4ClF3
180.55
MFCD00236319
JMXPOOVDUVHJRO-UHFFFAOYSA-N
2,4,5-trifluorobenzyl chloride, 1-chloromethyl-2,4,5-trifluorobenzene, 2,4,5-trifluorobenzylchloride, 2,4,5-trifluorobenzyl choride, benzene, 1-chloromethyl-2,4,5-trifluoro, alpha-chloro-2,4,5-trifluorotoluene, 1-chloromethyl-2,4,5-trifluoro-benzene, pubchem4024, acmc-1cjlx
2777063
1-(chloromethyl)-2,4,5-trifluorobenzene
FC1=CC(F)=C(CCl)C=C1F
This item is not returnable. View return policy
243139-71-1
C7H4ClF3
180.55
MFCD00236319
JMXPOOVDUVHJRO-UHFFFAOYSA-N
2,4,5-trifluorobenzyl chloride, 1-chloromethyl-2,4,5-trifluorobenzene, 2,4,5-trifluorobenzylchloride, 2,4,5-trifluorobenzyl choride, benzene, 1-chloromethyl-2,4,5-trifluoro, alpha-chloro-2,4,5-trifluorotoluene, 1-chloromethyl-2,4,5-trifluoro-benzene, pubchem4024, acmc-1cjlx
2777063
1-(chloromethyl)-2,4,5-trifluorobenzene
FC1=CC(F)=C(CCl)C=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.689
Flash Point 65°C (150°F)
Refractive Index 1.505
Quantity 1 g
UN Number UN3265
Formula Weight 180.56
Percent Purity 97%
Chemical Name or Material 2,4,5-Trifluorobenzyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'