missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzeneacetic Acid, TRC

Product Code. 30455984
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30455984 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455984 Supplier Toronto Research Chemicals Supplier No. TRCT7926251G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 209995-38-0
Molecular Formula C8H5F3O2
Quantity 1 g
Synonym (2,4,5-Trifluorophenyl)acetic Acid
InChI Formula InChI=1S/C8H5F3O2/c9-5-3-7(11)6(10)1-4(5)2-8(12)13/h1,3H,2H2,(H,12,13)
SMILES OC(=O)Cc1cc(F)c(F)cc1F
IUPAC Name 2-(2,4,5-trifluorophenyl)acetic acid
Molecular Weight (g/mol) 190.12
Formula Weight 190.0242
Chemical Name or Material 2,4,5-Trifluorobenzeneacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.