missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluoroaniline, 98%

Product Code. 11482778
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482778 5 g 5g
11492778 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482778 Supplier Thermo Scientific Alfa Aesar Supplier No. A16765.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 367-34-0 | C6H4F3N | 147.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-34-0
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.1
MDL Number MFCD00007649
InChI Key QMYVWJVVVMIBMM-UHFFFAOYSA-N
Synonym 2,4,5-trifluorophenylamine, benzenamine, 2,4,5-trifluoro, pubchem2469, 2,4,5-triflouroaniline, 2,4,5-trifloro aniline, acmc-1cs4m, 2,4,5-trifluoro-aniline, 2,4,5-trifluoro aniline, ksc497k9p, 2,4,5-trifluoro-phenylamine
PubChem CID 94953
IUPAC Name 2,4,5-trifluoroaniline
SMILES C1=C(C(=CC(=C1F)F)F)N

Specifications

Melting Point 59°C to 63°C
Quantity 5 g
UN Number UN2811
Beilstein 2085734
Formula Weight 147.1
Percent Purity 98%
Chemical Name or Material 2,4,5-Trifluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.