missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-trifluoro-3-methoxybenzoyl chloride, Thermo Scientific™

Product Code. 10163184
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10163184 25 g 25g
10756633 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163184 Supplier Thermo Scientific Maybridge Supplier No. SB02064EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 112811-66-2
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.563
InChI Key JVQSZTJTWSUJCR-UHFFFAOYSA-N
Synonym 3-methoxy-2,4,5-trifluorobenzoyl chloride, pubchem8384, acmc-1c4v9, 2,4,5-trifluoro-3-methoxy-benzoyl chloride, benzoyl chloride,2,4,5-trifluoro-3-methoxy, benzoyl chloride, 2,4,5-trifluoro-3-methoxy, 2,4,5-trifluoro-3-methoxybenzoyl chloride, 2,4,5-tris fluoranyl-3-methoxy-benzoyl chloride
PubChem CID 2775281
IUPAC Name 2,4,5-trifluoro-3-methoxybenzoyl chloride
SMILES COC1=C(C(=CC(=C1F)F)C(=O)Cl)F

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 25 g
Formula Weight 224.56
Percent Purity 97%
Chemical Name or Material 2, 4, 5-Trifluoro-3-methoxybenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.