missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Tribromoimidazole, 97%, Thermo Scientific™

Product Code. 10173790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173790 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10173790 Supplier Thermo Scientific Acros Supplier No. 161640050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2034-22-2
MDL Number MFCD00005184
InChI Key JCGGPCDDFXIVQB-UHFFFAOYSA-N
Synonym 2,4,5-tribromoimidazole, 1h-imidazole, 2,4,5-tribromo, imidazole, 2,4,5-tribromo, 2,4,5-tribromoimidazo, tribromoimidazole, pubchem7150, 2,5-tribromoimidazole, acmc-1cmbx, timtec-bb sbb001252, imidazole,4,5-tribromo
PubChem CID 16253
IUPAC Name 2,4,5-tribromo-1H-imidazole
SMILES C1(=C(N=C(N1)Br)Br)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 208.0°C to 212.0°C
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C3HBr3N2
Quantity 5 g
Beilstein 23, 49
Molecular Weight (g/mol) 304.76
Formula Weight 304.76
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 4, 5-Tribromoimidazole
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,5-Tribromoimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.