missing translation for 'onlineSavingsMsg'
Learn More

2-{4-[5-chloro-3-(trifluoromethyl)-2-pyridyl]piperazino}ethan-1-ol, 97%, Thermo Scientific™

Product Code. 10096172
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10096172 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10096172 Supplier Thermo Scientific Maybridge Supplier No. AW00098CB
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 215434-39-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H15ClF3N3O
MDL Number MFCD00203911
Quantity 250 mg
Synonym 2-4-5-chloro-3-trifluoromethyl-2-pyridyl piperazino ethan-1-ol, 2-4-5-chloro-3-trifluoromethyl pyridin-2-yl piperazin-1-yl ethanol, maybridge1_000177, 1-piperazineethanol,4-5-chloro-3-trifluoromethyl-2-pyridinyl, 2-4-5-chloro-3-trifluoromethyl pyridin-2-yl piperazin-1-yl ethan-1-ol
InChI Key NYMGNJNWMTZCFC-UHFFFAOYSA-N
SMILES C1CN(CCN1CCO)C2=NC=C(C=C2C(F)(F)F)Cl
IUPAC Name 2-[4-[5-chloro-3-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl]ethanol
Molecular Weight (g/mol) 309.717
PubChem CID 2795584
Formula Weight 309.72
Percent Purity 97%
Chemical Name or Material 2-{4-[5-Chloro-3-(trifluoromethyl)-2-pyridyl]piperazino}ethan-1-ol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.