missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetrahydro-1H-pyrazolo[3,4-b]pyridine, TRC

Product Code. 30407967
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407967 500 mg -
30406655 1 g -
30390729 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407967 Supplier Toronto Research Chemicals Supplier No. TRCH160031500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 933734-43-1
Molecular Formula C6H9N3
Molecular Weight (g/mol) 123.16
Synonym 1H,4H,5H,6H,7H-Pyrazolo[3,4-b]pyridine
IUPAC Name 4,5,6,7-tetrahydro-2H-pyrazolo[3,4-b]pyridine
SMILES C1CNc2n[nH]cc2C1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H9N3/c1-2-5-4-8-9-6(5)7-3-1/h4H,1-3H2,(H2,7,8,9)
Formula Weight 123.0796
Chemical Name or Material 2,4,5,6-Tetrahydro-1H-pyrazolo[3,4-b]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.