missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetrafluoropyrimidine, 95%

Product Code. 15485167
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485167 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15485167 Supplier Thermo Scientific Alfa Aesar Supplier No. H32453.MD
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 767-79-3 | C4F4N2 | 152.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 767-79-3
Boiling Point 83°C
Molecular Formula C4F4N2
MDL Number MFCD10687402
Quantity 250 mg
UN Number UN3265
Synonym tetrafluoropyrimidine, pyrimidine, tetrafluoro, pyrimidine,tetrafluoro, perfluoropyrimidine
InChI Key KZMWBUVUQLGBBP-UHFFFAOYSA-N
SMILES C1(=C(N=C(N=C1F)F)F)F
IUPAC Name 2,4,5,6-tetrafluoropyrimidine
Molecular Weight (g/mol) 152.052
PubChem CID 136605
Formula Weight 152.05
Percent Purity 95%
Chemical Name or Material 2,4,5,6-Tetrafluoropyrimidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.