missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetrachloropyrimidine, 97%

Product Code. 10357711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10357711 5 g 5g
15269826 1 g 1g
10728172 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10357711 Supplier Thermo Scientific Acros Supplier No. 333520050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1780-40-1 | C4Cl4N2 | 217.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1780-40-1
Molecular Formula C4Cl4N2
Molecular Weight (g/mol) 217.87
MDL Number MFCD00006062
InChI Key GVBHCMNXRKOJRH-UHFFFAOYSA-N
Synonym perchloropyrimidine, pyrimidine, tetrachloro, tetrachloropyrimidine, pyrimidine, 2,4,5,6-tetrachloro, 2,4,5,6-tetrachloro-1,3-pyrimidine, tetrachlorpyrimidin, tetrachloro-pyrimidin, acmc-1bsuq, 2,5,6-tetrachloropyrimidine, 5-23-05-00347 beilstein handbook reference
PubChem CID 15690
IUPAC Name 2,4,5,6-tetrachloropyrimidine
SMILES C1(=C(N=C(N=C1Cl)Cl)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 67°C to 68.5°C
Color White
Boiling Point 108°C to 112°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 217.87
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5,6-Tetrachloropyrimidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.