missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetraaminopyrimidine Sulfate, TRC

Product Code. 30425154
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425154 25 g -
30421999 250 g 250g
30424116 100 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425154 Supplier Toronto Research Chemicals Supplier No. TRCT28394025G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5392-28-9
Molecular Formula C4 H8 N6 . H2 O4 S
Molecular Weight (g/mol) 238.225
Synonym Pyrimidine, tetraamino-, sulfate (1:1) (8CI), Pyrimidinetetramine, sulfate (1:1) (9CI), 2,4,5,6-Tetraaminopyrimidine sulfate (1:1), NSC 227862, NSC 3277
IUPAC Name pyrimidine-2,4,5,6-tetramine;sulfuric acid
SMILES Nc1nc(N)c(N)c(N)n1.OS(=O)(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H8N6.H2O4S/c5-1-2(6)9-4(8)10-3(1)7;1-5(2,3)4/h5H2,(H6,6,7,8,9,10);(H2,1,2,3,4)
Formula Weight 238.0484
Chemical Name or Material 2,4,5,6-Tetraaminopyrimidine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.