missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetraaminopyrimidine sulfate, Dr. Ehrenstorfer

Product Code. 30325951
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325951 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325951 Supplier Dr. Ehrenstorfer Supplier No. DREC17323000
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,5,6-Tetraaminopyrimidine Sulfate
CAS 5392-28-9
Analyte or Component Names 2,4,5,6-Tetraaminopyrimidine Sulfate
Shipping Condition Room Temperature
Molecular Formula C4 H8 N6 . H2 O4 S
Certifications/Compliance ISO/IEC 17025
Synonym Pyrimidine, tetraamino-, sulfate (1:1) (8CI), Pyrimidinetetramine, sulfate (1:1) (9CI), 2,4,5,6-Tetraaminopyrimidine sulfate (1:1), NSC 227862, NSC 3277
InChI Formula InChI=1S/C4H8N6.H2O4S/c5-1-2(6)9-4(8)10-3(1)7;1-5(2,3)4/h5H2,(H6,6,7,8,9,10);(H2,1,2,3,4)
Solution Type Neat
SMILES Nc1nc(N)c(N)c(N)n1.OS(=O)(=O)O
IUPAC Name pyrimidine-2,4,5,6-tetramine;sulfuric acid
Industry Type Environmental Testing
Molecular Weight (g/mol) 238.22
Quantity 100 mg
Formula Weight 238.0484 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.