missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetraaminopyrimidine sulfate hydrate, 98%, Thermo Scientific Chemicals

Product Code. 10133570
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5392-28-9
Molecular Formula C4H10N6O4S
Molecular Weight (g/mol) 238.22
MDL Number MFCD06408008
InChI Key MQEFDQWUCTUJCP-UHFFFAOYSA-N
Synonym 2,4,5,6-tetraaminopyrimidine sulfate, pyrimidinetetramine sulfate, pyrimidine-2,4,5,6-tetraamine sulfate, pyrimidine-2,4,5,6-tetramine; sulfuric acid, tetraaminopyrimidine sulfate, pyrimidinetetramine, sulfate 1:1, pyrimidinetetramine, sulfate, tetraaminopyrimidine sulphate, unii-53qqy0i99m, tetra-aminopyrimidine sulfate
PubChem CID 79358
SMILES OS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Ochre to Brown
Loss on Drying 9% max. (1 g, 150°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 25, 423
Solubility Information Solubility in water: slightly soluble
IUPAC Name pyrimidine-2,4,5,6-tetramine; sulfuric acid
Formula Weight 238.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5,6-Tetraaminopyrimidine sulfate hydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,5,6-Tetraaminopyrimidine sulfate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'