missing translation for 'onlineSavingsMsg'
Learn More

2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid

Product Code. 15430367
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430367 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15430367 Supplier Thermo Scientific Alfa Aesar Supplier No. J64414.MA
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 19231-60-8 | C10H3I4NO4 | 708.755 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 19231-60-8
Molecular Formula C10H3I4NO4
MDL Number MFCD05023065
Quantity 10 mg
Synonym n,n-tetraiodo-phthaloyl-glycine, 2-4,5,6,7-tetraiodo-1,3-dioxoisoindol-2-yl acetic acid, 2-4,5,6,7-tetraiodo-1,3-dioxoisoindolin-2-yl acetic acid, 4,5,6,7-tetraiodo-1,3-dioxoisoindol-2-yl acetic acid, 4,5,6,7-tetraiodo-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-4,5,6,7-tetraiodo-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid
InChI Key FLSXICNYHDJIAW-UHFFFAOYSA-N
SMILES C(C(=O)O)N1C(=O)C2=C(C1=O)C(=C(C(=C2I)I)I)I
IUPAC Name 2-(4,5,6,7-tetraiodo-1,3-dioxoisoindol-2-yl)acetic acid
Molecular Weight (g/mol) 708.755
PubChem CID 4462449
Formula Weight 708.75
Physical Form Powder
Chemical Name or Material 2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.