missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethyl-2-pentene, 97%

Product Code. 11475656
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11475656

Brand: Thermo Scientific Alfa Aesar B20773.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-40-4 | C8H16 | 112.216 g/mol

2,4,4-Trimethyl-2-pentene was used to study the kinetics of liquid-phase hydrogenation of isooctenes on Pd/γ-Al2O3 catalyst in semibatch reactor. It was used in the analysis of butene oligomerate produced over solid and liquid phosphoric acid by GC-MS techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,4-Trimethyl-2-pentene was used to study the kinetics of liquid-phase hydrogenation of isooctenes on Pd/γ-Al2O3 catalyst in semibatch reactor. It was used in the analysis of butene oligomerate produced over solid and liquid phosphoric acid by GC-MS techniques.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-40-4
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00008902
InChI Key LAAVYEUJEMRIGF-UHFFFAOYSA-N
Synonym 2,4,4-trimethyl-2-pentene, 2-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-3-pentene, 2,4,4-trimethylpentene-2, unii-548r0du21i, beta-diisobutylene, propene, 1-tert-butyl-2-methyl, acmc-1cujo, dsstox_cid_6766, dsstox_rid_78208
PubChem CID 7869
IUPAC Name 2,4,4-trimethylpent-2-ene
SMILES CC(=CC(C)(C)C)C

Specifications

Melting Point -106°C
Density 0.72
Boiling Point 103°C to 105°C
Flash Point −1°C (29°F)
Refractive Index 1.416
Quantity 100 g
UN Number UN3295
Solubility Information Insoluble in water.
Formula Weight 112.22
Percent Purity 97%
Chemical Name or Material 2,4,4-Trimethyl-2-pentene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.