missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic Acid (Fenofibric Acid), Mikromol™

Product Code. 30049756
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049756 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049756 Supplier Mikromol Supplier No. MM0505.020025
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic Acid (Fenofibric Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenofibric Acid
CAS 42017-89-0
Analyte or Component Names Fenofibric Acid
Shipping Condition Room Temperature
Molecular Formula C17 H15 Cl O4
Certifications/Compliance ISO 17025
Synonym Fenofibrate Imp. B (EP), Fenofibrate USP Related Compound B, Fenofibrate USP RC B, 2[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid, Fenofibric Acid, Propanoic acid, 2-[4-(4-chlorobenzoyl)phenoxy]-2-methyl-, 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid, 2-[4'-(p-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, 2-[p-(p-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, FNF acid, LF 153, LF 178 acid, NSC 281318, Procetofenic acid, alpha-1081, Fenofibrate Related Compound B, Fenofibrate Impurity B
InChI Formula InChI=1S/C17H15ClO4/c1-17(2,16(20)21)22-14-9-5-12(6-10-14)15(19)11-3-7-13(18)8-4-11/h3-10H,1-2H3,(H,20,21)
SMILES CC(C)(Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2)C(=O)O
IUPAC Name 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid
Molecular Weight (g/mol) 318.75
Quantity 25 mg
Formula Weight 318.0659 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.