missing translation for 'onlineSavingsMsg'
Learn More

2-[4-[3-(10H-Phenothiazin-10-yl)propyl]piperazin-1-yl]ethanol, Mikromol™

Product Code. 30050753
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30050753

Brand: Mikromol MM0826.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[4-[3-(10H-Phenothiazin-10-yl)propyl]piperazin-1-yl]ethanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[4-[3-(10H-Phenothiazin-10-yl)propyl]piperazin-1-yl]ethanol
CAS 3533-97-9
Analyte or Component Names 2-[4-[3-(10H-Phenothiazin-10-yl)propyl]piperazin-1-yl]ethanol
Shipping Condition Room Temperature
Molecular Formula C21 H27 N3 O S
Certifications/Compliance ISO 17025
Synonym Perphenazine Imp. B (EP), Perphenazine USP Related Compound B, Perphenazine USP RC B, 2-[4-[3-(10H-Phenothiazin-10-yl)propyl]piperazin-1-yl]ethanol, Perphenazine Related Compound B, Perphenazine Impurity B
InChI Formula InChI=1S/C21H27N3OS/c25-17-16-23-14-12-22(13-15-23)10-5-11-24-18-6-1-3-8-20(18)26-21-9-4-2-7-19(21)24/h1-4,6-9,25H,5,10-17H2
SMILES OCCN1CCN(CCCN2c3ccccc3Sc4ccccc24)CC1
IUPAC Name 2-[4-(3-phenothiazin-10-ylpropyl)piperazin-1-yl]ethanol
Molecular Weight (g/mol) 369.52
Quantity 25 mg
Formula Weight 369.1875 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.