missing translation for 'onlineSavingsMsg'
Learn More

2-[4-[(2-Butyl-4-chloro-5-formylimidazol-1-yl)methyl]phenyl]benzonitrile, Mikromol™

Product Code. 30051631
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051631 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051631 Supplier Mikromol Supplier No. MM0168.380025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[4-[(2-Butyl-4-chloro-5-formylimidazol-1-yl)methyl]phenyl]benzonitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[4-[(2-Butyl-4-chloro-5-formylimidazol-1-yl)methyl]phenyl]benzonitrile
CAS 124750-67-0
Analyte or Component Names 2-[4-[(2-Butyl-4-chloro-5-formylimidazol-1-yl)methyl]phenyl]benzonitrile
Shipping Condition Room Temperature
Molecular Formula C22 H20 Cl N3 O
Certifications/Compliance ISO 9001
Synonym [1,1'-Biphenyl]-2-carbonitrile, 4'-[(2-butyl-4-chloro-5-formyl-1H-imidazol-1-yl)methyl]-, 4'-[(2-Butyl-4-chloro-5-formyl-1H-imidazol-1-yl)methyl][1,1'-biphenyl]-2-carbonitrile, 2-[4-[(2-Butyl-4-chloro-5-formylimidazol-1-yl)methyl]phenyl]benzonitrile, 4'-(2-Butyl-4-chloro-5-formylimidazol-1-ylmethyl)biphenyl-2-carbonitrile
InChI Formula InChI=1S/C22H20ClN3O/c1-2-3-8-21-25-22(23)20(15-27)26(21)14-16-9-11-17(12-10-16)19-7-5-4-6-18(19)13-24/h4-7,9-12,15H,2-3,8,14H2,1H3
SMILES CCCCc1nc(Cl)c(C=O)n1Cc2ccc(cc2)c3ccccc3C#N
IUPAC Name 2-[4-[(2-butyl-4-chloro-5-formylimidazol-1-yl)methyl]phenyl]benzonitrile
Molecular Weight (g/mol) 377.8667
Quantity 25 mg
Formula Weight 377.1295 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.