missing translation for 'onlineSavingsMsg'
Learn More

2-(4-(2-(4-(Carboxymethoxy)-2,3-dichlorobenzoyl)-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl)-2,3-dichlorophenoxy)acetic Acid, TRC

Product Code. 30367046
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367046 100 mg 100mg
30350964 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30367046 Supplier Toronto Research Chemicals Supplier No. TRCC178680100MG
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25355-92-4
Molecular Formula C26 H24 Cl4 O8
Molecular Weight (g/mol) 606.28
Synonym Acetic acid, [4-[2-[4-(carboxymethoxy)-2,3-dichlorobenzoyl]-3,4-dihydro-2,5-diethyl-2H-pyran-6-yl]-2,3-dichlorophenoxy]- (9CI), Acetic acid, [4-[2-(α-carboxy-2,3-dichloro-p-anisoyl)-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl]-2,3-dichlorophenoxy]- (8CI), 2-(4-(2-(4-(Carboxymethoxy)-2,3-dichlorobenzoyl)-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl)-2,3-dichlorophenoxy)acetic acid, [4-[2-[4-(Carboxymethoxy)-2,3-dichlorobenzoyl]-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl]-2,3-dichlorophenoxy]acetic acid
IUPAC Name 2-[4-[2-[4-(carboxymethoxy)-2,3-dichlorobenzoyl]-2,5-diethyl-3,4-dihydropyran-6-yl]-2,3-dichlorophenoxy]acetic acid
SMILES CCC1=C(OC(CC)(CC1)C(=O)c2ccc(OCC(=O)O)c(Cl)c2Cl)c3ccc(OCC(=O)O)c(Cl)c3Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C26H24Cl4O8/c1-3-13-9-10-26(4-2,25(35)15-6-8-17(23(30)21(15)28)37-12-19(33)34)38-24(13)14-5-7-16(22(29)20(14)27)36-11-18(31)32/h5-8H,3-4,9-12H2,1-2H3,(H,31,32)(H,33,34)
Formula Weight 604.0225
Chemical Name or Material 2-(4-(2-(4-(Carboxymethoxy)-2,3-dichlorobenzoyl)-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl)-2,3-dichlorophenoxy)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.