missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[[4-[(1E)-2-[4-[[2-(2,5-Dihydro-2,5-dioxo-1H-pyrrol-1-yl)acetyl]amino]phenyl]diazenyl]phenyl]amino]-N,N,N-triethyl-2-oxoethanaminium Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-((4-((1E)-2-(4-((2-(2,5-Dihydro-2,5-dioxo-1H-pyrrol-1-yl)acetyl)amino)phenyl)diazenyl)phenyl)amino)-N,N,N-triethyl-2-oxoethanaminium Chloride |
| Analyte or Component Names | 2-((4-((1E)-2-(4-((2-(2,5-Dihydro-2,5-dioxo-1H-pyrrol-1-yl)acetyl)amino)phenyl)diazenyl)phenyl)amino)-N,N,N-triethyl-2-oxoethanaminium Chloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H31 N6 O4 . Cl |
| InChI Formula | InChI=1S/C26H30N6O4.ClH/c1-4-32(5-2,6-3)18-24(34)28-20-9-13-22(14-10-20)30-29-21-11-7-19(8-12-21)27-23(33)17-31-25(35)15-16-26(31)36;/h7-16H,4-6,17-18H2,1-3H3,(H-,27,28,29,30,33,34,35,36);1H |
| Solution Type | Neat |
| SMILES | [Cl-].CC[N+](CC)(CC)CC(=O)Nc1ccc(cc1)\N=N\c2ccc(NC(=O)CN3C(=O)C=CC3=O)cc2 |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 527.015 |
| Quantity | 25 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?