missing translation for 'onlineSavingsMsg'
Learn More

2-(4-(1-Hydroxy-4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butyl)phenyl)propanoic Acid Hydrochloride, TRC

Product Code. 30393924
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393924 50 mg 50mg
30389681 25 mg -
30399208 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393924 Supplier Toronto Research Chemicals Supplier No. TRCH95404350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C31H37NO4 . HCl
Molecular Weight (g/mol) 487.6
Synonym Fexofenadine Impurity F
SMILES CC(C(=O)O)c1ccc(cc1)C(O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C31H37NO4/c1-23(30(34)35)24-14-16-25(17-15-24)29(33)13-8-20-32-21-18-28(19-22-32)31(36,26-9-4-2-5-10-26)27-11-6-3-7-12-27/h2-7,9-12,14-17,23,28-29,33,36H,8,13,18-22H2,1H3,(H,34,35)
Chemical Name or Material 2-(4-(1-Hydroxy-4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butyl)phenyl)propanoic Acid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.