missing translation for 'onlineSavingsMsg'
Learn More

α-2,3-sialyltransferase-IN-1, MedChemExpress

Product Code. 16497357
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497357 5 mg 5mg
16450937 50 mg 50mg
16424648 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497357 Supplier Medchem Express Supplier No. HY112535
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress α-2,3-sialyltransferase-IN-1 (Lith-O-Asp analog) is a noncompetitive α-2,3-sialyltransferase inhibitor with an IC50 of 6 μM.

α-2,3-sialyltransferase-IN-1 (Lith-O-Asp analog) is a noncompetitive α-2,3-sialyltransferase inhibitor with an IC50 of 6 μM.

Specifications

Chemical Name or Material α-2,3-sialyltransferase-IN-1
Physical Form Solid
CAS 881179-06-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C28H45NO6
For Use With (Application) Neuroscience-Neuromodulation
Synonym Lith-O-Asp analog
Solubility Information DMSO : ≥ 103.7 mg/mL (210.92 mM)
SMILES C[C@@]1([C@@]2([H])[C@H](C)CCC(O)=O)[C@](CC2)([H])[C@@](CC[C@@]3([H])[C@@]4(CC[C@@H](OC([C@H](N)CC(O)=O)=O)C3)C)([H])[C@]4([H])CC1
Molecular Weight (g/mol) 491.66
Formula Weight 491.66
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.