missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Pyridyl)benzimidazole-6-carboxylic acid, 97%

Product Code. 15421376
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421376 250 mg 250mg
15470848 1 g 1g
15480848 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421376 Supplier Thermo Scientific Alfa Aesar Supplier No. H54593.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 312496-16-5 | C13H9N3O2 | 239.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312496-16-5
Molecular Formula C13H9N3O2
Molecular Weight (g/mol) 239.234
MDL Number MFCD01414693
InChI Key DHRPCXYIVJXXJR-UHFFFAOYSA-N
Synonym 1h-benzimidazole-6-carboxylicacid, 2-3-pyridinyl, 2-pyridin-3-yl-3h-1,3-benzodiazole-5-carboxylic acid, 2-pyridin-3-yl-3h-benzoimidazole-5-carboxylic acid, 2-3-pyridyl benzimidazole-6-carboxylic acid, 6-carboxy-2-pyridin-3-yl-1h-benzimidazole, 2-pyridin-3-yl-1h-benzoimidazole-5-carboxylic acid, 2-3-pyridinyl-1h-benzoimidazole-5-carboxylic acid, 2-pyridin-3-yl-1h-benzimidazole-5-carboxylic acid, 2-pyridin-3-yl-3h-benzimidazole-5-carboxylic acid
PubChem CID 2771805
IUPAC Name 2-pyridin-3-yl-3H-benzimidazole-5-carboxylic acid
SMILES C1=CC(=CN=C1)C2=NC3=C(N2)C=C(C=C3)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 239.23
Percent Purity 97%
Chemical Name or Material 2-(3-Pyridyl)benzimidazole-6-carboxylic acid
compliance-icons
Product Identifier
  • 2-(3-Pyridyl)benzimidazole-6-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-(3-Pyridyl)benzimidazole-6-carboxylicacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.