missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Piperidinyloxy)-N-propylacetamide

Product Code. 11489770
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11489770

Brand: Alfa Aesar™ H50994.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 902837-33-6
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD08064279
InChI Key HIPDESKOLOAVFF-UHFFFAOYSA-N
Synonym 2-piperidin-3-yloxy-n-propylacetamide, 2-3-piperidyloxy-n-propylacetamide, 2-piperidin-3-yl oxy-n-propylacetamide, 2-3-piperidinyloxy-n-propylacetamide
PubChem CID 43196816
IUPAC Name 2-piperidin-3-yloxy-N-propylacetamide
SMILES CCCNC(=O)COC1CCCNC1

Specifications

Quantity 1 g
Formula Weight 200.28
Chemical Name or Material 2-(3-Piperidinyloxy)-N-propylacetamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.