missing translation for 'onlineSavingsMsg'
Learn More

2',3'-O-Isopropylideneuridine, TRC

Product Code. 30399337
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399337 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399337 Supplier Toronto Research Chemicals Supplier No. TRCI86900025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 362-43-6
Molecular Formula C12 H16 N2 O6
Quantity 25 g
InChI Formula InChI=1S/C12H16N2O6/c1-12(2)19-8-6(5-15)18-10(9(8)20-12)14-4-3-7(16)13-11(14)17/h3-4,6,8-10,15H,5H2,1-2H3,(H,13,16,17)/t6-,8-,9-,10-/m1/s1
SMILES CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N3C=CC(=O)NC3=O
IUPAC Name 1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]pyrimidine-2,4-dione
Molecular Weight (g/mol) 284.27
Formula Weight 284.1008
Chemical Name or Material 2',3'-O-Isopropylideneuridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.