missing translation for 'onlineSavingsMsg'
Learn More

2',3'-O-Isopropylideneuridine, 99%, Thermo Scientific™

Product Code. 10775571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10775571 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10775571 Supplier Thermo Scientific Acros Supplier No. 122740010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 362-43-6
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C12H16N2O6
MDL Number MFCD00034509
Quantity 1g
Synonym 2',3'-isopropylideneuridine, 2',3'-o-isopropylidene uridine, 2',3'-o-isopropylideneuridine, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4 1h,3h-dione, 1-1r,2r,4r,5r-4-hydroxymethyl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oct-2-yl-1,3-dihydropyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl-3h-pyrimidine-2,4-dione, 2/',3/'-isopropylideneuridine, 2',3'-o-isopropylideneuridine hplc, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl-1,2,3,4-tetrahydropyrimidine-2,4-dione
InChI Key GFDUSNQQMOENLR-PEBGCTIMSA-N
SMILES CC1(OC2C(OC(C2O1)N3C=CC(=O)NC3=O)CO)C
IUPAC Name 1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]pyrimidine-2,4-dione
Molecular Weight (g/mol) 284.268
PubChem CID 64967
Formula Weight 284.26
Percent Purity 99%
Chemical Name or Material 2', 3'-O-Isopropylideneuridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.