missing translation for 'onlineSavingsMsg'
Learn More

2,3-O-Isopropylidene-D-glyceraldehyde, 50% w/w in dichloromethane

Product Code. 15470557
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470557 10 g 10g
15450557 1 g 1g
15460557 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15470557 Supplier Thermo Scientific Alfa Aesar Supplier No. J66337.09
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15186-48-8 | C6H10O3 | 130.143 g/mol

Reactant involved in organic reactions including, Henry condensation reactions, Synthesis of highly functionalized chiral aziridines, Olefination of aldehydes, C-H oxidations, Aldol-type condensations, Synthesis of hydrophilic pyrazine dyes for use as exogenous fluorescent tracer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant involved in organic reactions including, Henry condensation reactions, Synthesis of highly functionalized chiral aziridines, Olefination of aldehydes, C-H oxidations, Aldol-type condensations, Synthesis of hydrophilic pyrazine dyes for use as exogenous fluorescent tracer agents.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15186-48-8
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00269682
InChI Key YSGPYVWACGYQDJ-YFKPBYRVSA-N
Synonym r-+-2,2-dimethyl-1,3-dioxolane-4-carboxaldehyde, 4r-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, r-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, r-+-glyceraldehyde acetonide, 1,3-dioxolane-4-carboxaldehyde, 2,2-dimethyl-, r, acetone d-glyceraldehyde, +-2,2-dimethyl-1,3-dioxolane-4-carboxaldehyde, 1,3-dioxolane-4-carboxaldehyde, 2,2-dimethyl-, 4r, 4r-2,2-dimethyl-1,3-dioxolan-4-carbaldehyd, pubchem5767
PubChem CID 259712
IUPAC Name (4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde
SMILES CC1(OCC(O1)C=O)C

Specifications

Color Colorless to Yellow
Flash Point 62°C (144°F)
Refractive Index 1.454
Quantity 10 g
UN Number 1593
Solubility Information Difficult to mix.
Optical Rotation +53.8° (c=2 in Chloroform)
Formula Weight 130.14
Concentration or Composition (by Analyte or Components) 50% w/w in dichloromethane
Physical Form Liquid
Chemical Name or Material 2,3-O-Isopropylidene-D-glyceraldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.