missing translation for 'onlineSavingsMsg'
Learn More

2,3-O-Isopropylidene-D-erythronolactone, 98%, Thermo Scientific Chemicals

Product Code. 15488399
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15488399

Brand: Thermo Scientific Alfa Aesar J66871.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use in spiroannulated carbohydrate synthesis

It undergoes Aldol condensations with silyl ketene acetals.1 Employed in spiroannulated carbohydrate synthesis.2 Convergent syntheses of a hydroxylated indolizidine,3 carbohydrate substituted benzoquinones,4 and of the oxazole segment of calyculin5 have been accomplished using this chiral synthon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It undergoes Aldol condensations with silyl ketene acetals.1 Employed in spiroannulated carbohydrate synthesis.2 Convergent syntheses of a hydroxylated indolizidine,3 carbohydrate substituted benzoquinones,4 and of the oxazole segment of calyculin5 have been accomplished using this chiral synthon.

Solubility
Soluble in water

Notes
Store at room temperature. Incompatible with heat, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25581-41-3
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00134440
InChI Key WHPSMBYLYRPVGU-UHFFFAOYNA-N
Synonym 2,3-o-isopropylidene-d-erythronolactone, --2,3-o-isopropylidene-d-erythronolactone, 3ar,6ar-2,2-dimethyldihydrofuro 3,4-d 1,3 dioxol-4 3ah-one, 3ar,6ar-2,2-dimethyl-6,6a-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, 3ar,6ar-2,2-dimethyl-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, pubchem19725, 2,3-o-isopropylidene-d-erythronolactol, d-2,3-o-isopropylidene-erythronic acid, a-lactone, 2-o,3-o-isopropylidene-d-erythronic acid gamma-lactone
PubChem CID 386843
IUPAC Name (3aR,6aR)-2,2-dimethyl-6,6a-dihydro-3aH-furo[3,4-d][1,3]dioxol-4-one
SMILES CC1(C)OC2COC(=O)C2O1

Specifications

Melting Point 67°C to 69°C
Color White
Quantity 1 g
Beilstein 1282952
Solubility Information Soluble in water
Optical Rotation −118° (c=1 in Water)
Formula Weight 158.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3-O-Isopropylidene-D-erythronolactone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.