missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Methylphenoxy)ethanol, 98%, Thermo Scientific Chemicals

Product Code. 11351787
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11351787 10 g 10g
11341787 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351787 Supplier Thermo Scientific Alfa Aesar Supplier No. L04984.09
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13605-19-1 | Molecular Formula: C9H12O2 | Molecular Weight: 152.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13605-19-1
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00014035
InChI Key FDBXUXVQIOQYIX-UHFFFAOYSA-N
Synonym 2-3-methylphenoxy ethanol, 2-m-tolyloxy ethanol, ethanol, 2-3-methylphenoxy, ethylene glycol mono-m-tolyl ether, m-cresoxyethanol, 2-3-tolyloxy ethanol, ethanol, 2-m-tolyloxy, 2-3'-methylphenoxy ethanol, ethylene glycol m-cresyl ether, 2-3-methylphenoxy ethan-1-ol
PubChem CID 83611
IUPAC Name 2-(3-methylphenoxy)ethanol
SMILES CC1=CC(=CC=C1)OCCO

Specifications

Boiling Point 104°C to 105°C (3 mmHg)
Quantity 10 g
Beilstein 2247221
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2-(3-Methylphenoxy)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.