missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Methoxyallylidene)malonic acid dimethyl ester, 95%

Product Code. 11995121 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11995121 10 g Glass bottle 10g
10490911 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11995121 Supplier Thermo Scientific Acros Supplier No. 302090100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41530-32-9 | C9H12O5 | 200.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41530-32-9
Molecular Formula C9H12O5
Molecular Weight (g/mol) 200.19
InChI Key NOBNTZJTOBIOHL-GQCTYLIASA-N
Synonym 2-3-methoxyallylidene malonic acid dimethyl ester, dimethyl 2-3-methoxyallylidene malonate, 1,3-dimethyl 2-2e-3-methoxyprop-2-en-1-ylidene propanedioate, 2-3-methoxyallylidene malonic acid dimethyl ester, 96, dimethyl 2-e-3-methoxyprop-2-enylidene propanedioate, propanedioic acid, 3-methoxy-2-propenylidene-, dimethyl ester, dimethyl e-2-3-methoxyallylidene malonate, 3-methoxy-2-propenylidene malonic acid dimethyl ester, dimethyl 2-2e-3-methoxy-2-propenylidene malonate #
PubChem CID 5374172
IUPAC Name dimethyl 2-[(E)-3-methoxyprop-2-enylidene]propanedioate
SMILES COC=CC=C(C(=O)OC)C(=O)OC

Specifications

Color Green-Yellow
Boiling Point 120.0°C to 125.0°C (0.3 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5370 to 1.5390
Quantity 10 g
Formula Weight 200.19
Percent Purity 95%
Physical Form Low Melting Solid
Chemical Name or Material 2-(3-Methoxyallylidene)malonic acid dimethyl ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.