missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Isopropylideneuridine

Product Code. 15468819
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468819 25 g 25g
11312266 1 g 1g
11322266 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15468819 Supplier Thermo Scientific Alfa Aesar Supplier No. L03686.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 362-43-6 | C12H16N2O6 | 284.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 362-43-6
Molecular Formula C12H16N2O6
Molecular Weight (g/mol) 284.268
MDL Number MFCD00034509
InChI Key GFDUSNQQMOENLR-PEBGCTIMSA-N
Synonym 2',3'-isopropylideneuridine, 2',3'-o-isopropylidene uridine, 2',3'-o-isopropylideneuridine, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4 1h,3h-dione, 1-1r,2r,4r,5r-4-hydroxymethyl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oct-2-yl-1,3-dihydropyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl-3h-pyrimidine-2,4-dione, 2/',3/'-isopropylideneuridine, 2',3'-o-isopropylideneuridine hplc, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl pyrimidine-2,4-dione, 1-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 64967
IUPAC Name 1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]pyrimidine-2,4-dione
SMILES CC1(OC2C(OC(C2O1)N3C=CC(=O)NC3=O)CO)C

Specifications

Melting Point 165°C to 166°C
Quantity 25 g
Beilstein 32671
Formula Weight 284.27
Chemical Name or Material 2',3'-Isopropylideneuridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.