missing translation for 'onlineSavingsMsg'
Learn More

2’,3’-Isopropylidene Adenosine-13C5, TRC

Product Code. 30394546
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394546 5 mg 5mg
30392051 2.5 mg 2.5mg
30397715 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394546 Supplier Toronto Research Chemicals Supplier No. TRCI8246775MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2’,3’-Isopropylidene Adenosine-13C5
Molecular Formula C813C5H17N5O4
Quantity 5 mg
Synonym 2',3'-O-(1-Methylethylidene)adenosine-13C5, NSC 29413-13C5
InChI Formula InChI=1S/C13H17N5O4/c1-13(2)21-8-6(3-19)20-12(9(8)22-13)18-5-17-7-10(14)15-4-16-11(7)18/h4-6,8-9,12,19H,3H2,1-2H3,(H2,14,15,16)/t6-,8-,9-,12-/m1/s1/i3+1,6+1,8+1,9+1,12+1
SMILES O[13CH2][13C@@H]1[13C@H]([13C@H]([13C@H](N2C=NC3=C(N=CN=C32)N)O1)O4)OC4(C)C
IUPAC Name ((3aR,4R,6R,6aR)-6-(6-amino-9H-purin-9-yl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl-3a,4,6,6a-13C4)methanol-13C
Molecular Weight (g/mol) 312.27
Formula Weight 312.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.