missing translation for 'onlineSavingsMsg'
Learn More

2-(3-hydroxyphenyl)propanoic acid, TRC

Product Code. 30393161
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393161 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393161 Supplier Toronto Research Chemicals Supplier No. TRCH100610100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(3-hydroxyphenyl)propanoic acid
CAS 56911-50-3
Molecular Formula C9 H10 O3
Quantity 100 mg
InChI Formula InChI=1S/C9H10O3/c1-6(9(11)12)7-3-2-4-8(10)5-7/h2-6,10H,1H3,(H,11,12)
SMILES CC(C(=O)O)c1cccc(O)c1
IUPAC Name 2-(3-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol) 166.174
Formula Weight 166.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.