missing translation for 'onlineSavingsMsg'
Learn More

2-[3-[(Hydroxyimino)methyl]-4-(2-methylpropoxy)phenyl]-4-methyl-5-Thiazolecarboxylic Acid Ethyl Ester, TRC

Product Code. 30398025
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398025 100 mg
30408388 1 g
30394805 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398025 Supplier Toronto Research Chemicals Supplier No. TRCH296910100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271738-74-9
Molecular Formula C18H22N2O4S
Molecular Weight (g/mol) 362.44
Synonym Febuxostat Impurity 6
IUPAC Name ethyl 2-[3-[(E)-hydroxyiminomethyl]-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)c1sc(nc1C)c2ccc(OCC(C)C)c(\C=N\O)c2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H22N2O4S/c1-5-23-18(21)16-12(4)20-17(25-16)13-6-7-15(24-10-11(2)3)14(8-13)9-19-22/h6-9,11,22H,5,10H2,1-4H3/b19-9+
Formula Weight 362.13
Chemical Name or Material 2-[3-[(Hydroxyimino)methyl]-4-(2-methylpropoxy)phenyl]-4-methyl-5-Thiazolecarboxylic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.