missing translation for 'onlineSavingsMsg'
Learn More

2,3-Heptanedione, 97+%

Product Code. 11459426
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459426 100 g 100g
11449426 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459426 Supplier Thermo Scientific Alfa Aesar Supplier No. A19136.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 96-04-8 | C7H12O2 | 128.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-04-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00036550
InChI Key FJPGAMCQJNLTJC-UHFFFAOYSA-N
Synonym 2,3-heptanedione, acetyl valeryl, valerylacetyl, acetylvaleryl, acetyl pentanoyl, unii-dk55dde86p, dk55dde86p, benzil-related compound, 47, heptanedione, fema no. 2543
PubChem CID 60983
IUPAC Name heptane-2,3-dione
SMILES CCCCC(=O)C(=O)C

Specifications

Density 0.92
Boiling Point 64°C (18 mmHg)
Flash Point 40°C (104°F)
Refractive Index 1.416
Quantity 100 g
UN Number UN1224
Beilstein 1700989
Formula Weight 128.17
Percent Purity ≥97%
Chemical Name or Material 2,3-Heptanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.