missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Formyl-4-methoxyphenyl)-2-(1-hydroxycyclohexyl)acetonitrile, TRC

Product Code. 30402409
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402409 50 mg 50mg
30394870 10 mg 10mg
30401758 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402409 Supplier Toronto Research Chemicals Supplier No. TRCF70073550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(3-Formyl-4-methoxyphenyl)-2-(1-hydroxycyclohexyl)acetonitrile
Analyte or Component Names 2-(3-Formyl-4-methoxyphenyl)-2-(1-hydroxycyclohexyl)acetonitrile
Shipping Condition Room Temperature
Molecular Formula C11 H15 Br N2 O
InChI Formula InChI=1S/C11H15BrN2O/c1-2-3-8-13-14-11(15)9-4-6-10(12)7-5-9/h4-7,13H,2-3,8H2,1H3,(H,14,15)
Solution Type Neat
SMILES CCCCNNC(=O)c1ccc(Br)cc1
IUPAC Name 4-bromo-N'-butylbenzohydrazide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 271.154
Quantity 50 mg
Formula Weight 270.037
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.