missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Fluorophenylamino)benzoic acid, 98%, Thermo Scientific™

Product Code. 11380518
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380518 10 g 10g
11300528 2 g 2g
11390518 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11380518 Supplier Thermo Scientific Alfa Aesar Supplier No. 043852.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-59-1
Molecular Formula C13H10FNO2
Molecular Weight (g/mol) 231.226
MDL Number MFCD01675228
InChI Key HYZIDPAMLUKNIR-UHFFFAOYSA-N
Synonym n-3-fluorophenyl anthranilic acid, 2-3-fluorophenyl amino benzoic acid, anthranilic acid, n-m-fluorophenyl, benzoic acid, 2-3-fluorophenyl amino, n-3-fluorophenyl anthranilsaeure german, acide n-3-flurophenyl anthranilique french, 2-3-fluorophenylamino benzoic acid, n-3-fluorophenyl anthranilsaeure, acide n-3-flurophenyl anthranilique
PubChem CID 120158
IUPAC Name 2-(3-fluoroanilino)benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)NC2=CC(=CC=C2)F

Specifications

Melting Point 158°C to 163°C
Color Gray to Yellow
Quantity 10 g
Formula Weight 231.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2- (3-Fluorophenylamino)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.