missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylthiophene, TRC

Product Code. 30388478
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388478 50 mg
30370366 500 mg
30372393 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388478 Supplier Toronto Research Chemicals Supplier No. TRCD46163350MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-16-6
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
SMILES Cc1ccsc1C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H8S/c1-5-3-4-7-6(5)2/h3-4H,1-2H3
Formula Weight 112.0347
Chemical Name or Material 2,3-Dimethylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.