Learn More
2,3-Dimethylthiophene, 97%
Description
It has also been found that 2,3-dimethylthiophene can be formylated to yield a dimethylthiophene aldehyde, which as expected proved to have the aldehyde group in the free a-position.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It has also been found that 2,3-dimethylthiophene can be formylated to yield a dimethylthiophene aldehyde, which as expected proved to have the aldehyde group in the free a-position.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Specifications
Specifications
| Boiling Point | 142°C |
| Flash Point | 26° (79°F) |
| Quantity | 5 g |
| UN Number | UN1993 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 112.19 |
| Percent Purity | 97% |
| Physical Form | Liquid |
| Chemical Name or Material | 2,3-Dimethylthiophene |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.