missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylquinoxaline, 97%

Product Code. 10020860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10020860 25 g 25g
10214050 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10020860 Supplier Thermo Scientific Acros Supplier No. 116780250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2379-55-7 | C10H10N2 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2379-55-7
MDL Number MFCD00006728
InChI Key FKHNZQFCDGOQGV-UHFFFAOYSA-N
PubChem CID 16925
IUPAC Name 2,3-dimethylquinoxaline
SMILES CC1=NC2=CC=CC=C2N=C1C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 102.0°C to 107.0°C
Color Beige
Boiling Point 257.0°C to 259.0°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C10H10N2
Quantity 25 g
Beilstein 23, 191
Synonym quinoxaline, 2,3-dimethyl, aurora ka-3449, quinoxaline,3-dimethyl, 2,3-dimethyl-quinoxalin, 2,3-dimethyl-quinoxaline, acmc-1cg5s, quinoxaline,2,3-dimethyl, salor-int l498777-1ea, 5-23-07-00267 beilstein handbook reference, chembl67865
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 158.2
Formula Weight 158.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Dimethylquinoxaline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dimethylquinoxaline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.