missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylphenylhydrazine hydrochloride, 97%

Product Code. 11427857
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427857 25 g 25g
11417857 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427857 Supplier Thermo Scientific Alfa Aesar Supplier No. A16526.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 56737-75-8 | C8H13ClN2 | 172.656 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56737-75-8
Molecular Formula C8H13ClN2
Molecular Weight (g/mol) 172.656
MDL Number MFCD00013380
InChI Key UKAYNIJBIXNDDJ-UHFFFAOYSA-N
Synonym 2,3-dimethylphenylhydrazine hydrochloride, 2,3-dimethylphenyl hydrazine hydrochloride, 1-2,3-dimethylphenyl hydrazine hydrochloride, acmc-1atp8, 3-hydrazino-o-xylene hydrochloride, 2,3-dimethylphenylhydrazine, chloride, hydrazine, 2,3-dimethylphenyl-, hydrochloride 1:1
PubChem CID 2774885
IUPAC Name (2,3-dimethylphenyl)hydrazine;hydrochloride
SMILES CC1=C(C(=CC=C1)NN)C.Cl

Specifications

Melting Point ∼210°C (decomposition)
Quantity 25 g
Beilstein 6096287
Formula Weight 172.66
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylphenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.