missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylphenylboronic acid, 97%

Product Code. 10186277 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10186277 5 g Glass bottle 5g
10708288 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10186277 Supplier Thermo Scientific Acros Supplier No. 393790050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 183158-34-1 | C8H11BO2 | 149.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183158-34-1
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.99
MDL Number MFCD01863524
InChI Key ZYYANAWVBDFAHY-UHFFFAOYSA-N
Synonym 2,3-dimethylbenzeneboronic acid, 2,3-dimethylphenyl boronic acid, o-xylene-3-boronic acid, 2,3-dimethylphenyl boranediol, 2,3-dimethylbenzeneboronicacid, 2,3-dimethylphenylboronicacid, pubchem5037, acmc-1bxk2, amtb394, ksc174k9l
PubChem CID 2773395
IUPAC Name (2,3-dimethylphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)C)C)(O)O

Specifications

Melting Point 175.0°C
CAS Min % 96.0
Color White
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 96% min. (HPLC)
Formula Weight 149.99
Percent Purity 97%
Chemical Name or Material 2, 3-Dimethylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dimethylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.