missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylpentane, 97%

Product Code. 11330308
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11330308 25 g 25g
11490600 1 g 1g
11340308 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11330308 Supplier Thermo Scientific Alfa Aesar Supplier No. 043707.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 565-59-3 | C7H16 | 100.21 g/mol

It is used in the liquid-phase oxidation of 2,4-dimethylpentane and in a study on the preparation and the physical constants of a number of alkanes and cycloalkanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-59-3
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009337
InChI Key WGECXQBGLLYSFP-UHFFFAOYNA-N
Synonym pentane, 2,3-dimethyl, 3,4-dimethylpentane, acmc-1an9i, 3-01-00-00445 beilstein handbook reference, 2,3-dimethylpentane, 2,3-dimethylpentane, analytical standard
PubChem CID 11260
IUPAC Name 2,3-dimethylpentane
SMILES CCC(C)C(C)C

Specifications

Density 0.690 g/mL
Boiling Point 89°C to 90°C
Flash Point −6°C (21°F)
Refractive Index 1.392
Quantity 25 g
UN Number UN1206
Solubility Information Insoluble in water.
Formula Weight 100.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylpentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.