missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylmaleic Anhydride, TRC

Product Code. 30377921
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377921 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377921 Supplier Toronto Research Chemicals Supplier No. TRCD4745801G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dimethylmaleic Anhydride
CAS 766-39-2
Molecular Formula C6H6O3
Quantity 1 g
Synonym Dimethyl-maleic anhydride, 2,3-dimethylmalonic anhydride4,4-Dimethyl-2,5-furandion, Dimethylmaleic acid anhydride, Dimethylmaleic Anhydride, NSC 92512
InChI Formula InChI=1S/C6H6O3/c1-3-4(2)6(8)9-5(3)7/h1-2H3
SMILES CC1=C(C)C(=O)OC1=O
Molecular Weight (g/mol) 126.11
Formula Weight 126.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.